Learn More
Waters Corp NanoEase M/Z Peptide BEH C18 Column, 300Å Pore/1.7 μm Particle, 100 μm × 100 mm
Catalog No. 502392370 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for nano- and micro-scale LC-MS applications, which are critical for samples with limited availability, requiring high sensitivity and low detection limits. They are compatible with ZenFit Connection Technology to ensure reliable, high-performance connections.
  • Designed for use with ZenFit Connection Technology.
  • ZenFit Connection Technology provides finger-tight re-usable connectors.
  • Prevents poor connections and maintains flow path integrity.
  • Reduces chromatographic variability.
  • Withstands pressures up to 15,000 psi.
  • Tolerates temperatures up to 90 °C.
  • Enhances sensitivity of mass spectrometry by increasing sampling efficiency.
  • Improves ionization efficiency and reduces matrix effects for low flow LC-MS.
Catalog No. 50-239-2370 Supplier Waters Corp Supplier No. 186008802
These columns are designed for nano- and micro-scale LC-MS applications, which are critical for samples with limited availability, requiring high sensitivity and low detection limits. They are compatible with ZenFit Connection Technology to ensure reliable, high-performance connections.
  • Designed for use with ZenFit Connection Technology.
  • ZenFit Connection Technology provides finger-tight re-usable connectors.
  • Prevents poor connections and maintains flow path integrity.
  • Reduces chromatographic variability.
  • Withstands pressures up to 15,000 psi.
  • Tolerates temperatures up to 90 °C.
  • Enhances sensitivity of mass spectrometry by increasing sampling efficiency.
  • Improves ionization efficiency and reduces matrix effects for low flow LC-MS.