Learn More
Waters Corp Column, nanoEase M/Z Peptide BEH C18, 300Å, 1.7 μm
Catalog No. 502391049 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility crucial for biomarker discovery, protein identification, and characterization. They are built to withstand operating pressures of up to 15kpsi and incorporate patented BEH Technology for robust performance across a wide pH range.
  • Delivers high sensitivity and resolution for protein identification.
  • Withstands up to 15kpsi operating pressure for longevity.
  • Features 300Å pore, C18, 1.7 μm BEH particles for wide pH stability and ultra-low bleed.
  • Includes ZenFit connectors for reliable, leak-free connections.
  • Manufactured in a cGMP, ISO 9001-certified facility, with comprehensive quality control.
Catalog No. 50-239-1049 Supplier Waters Corp Supplier No. 186008801
These columns are designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility crucial for biomarker discovery, protein identification, and characterization. They are built to withstand operating pressures of up to 15kpsi and incorporate patented BEH Technology for robust performance across a wide pH range.
  • Delivers high sensitivity and resolution for protein identification.
  • Withstands up to 15kpsi operating pressure for longevity.
  • Features 300Å pore, C18, 1.7 μm BEH particles for wide pH stability and ultra-low bleed.
  • Includes ZenFit connectors for reliable, leak-free connections.
  • Manufactured in a cGMP, ISO 9001-certified facility, with comprehensive quality control.