Learn More
Waters Corp Waters nanoEase M/Z Peptide BEH C18 Column, 130Å, 1.7 μm, 75 μm X 150 mm
Supplier:  Waters Corp 186008793
Encompass_Preferred
These columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, providing high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification and characterization. They are designed to withstand 15 kpsi operating pressure and manage pressure fluctuations during injection, contributing to superior longevity.
  • ZenFit connectors provide easy-to-use, fingertight, and tool-free liquid connections for leak-free performance.
  • Utilizes patented BEH Technology with 130Å pore, C18, and 1.7 μm particles.
  • Offers a broad usable pH range (1 to 10 pH) and excellent low pH stability.
  • Suitable for proteomics, protein characterization, and peptide synthesis.
  • Each batch undergoes rigorous QC testing for peptide separation.
  • Ultra-low dispersion design for minimized peak tailing and optimal efficiency.
Catalog No. 50-203-8815