Learn More
Waters Corp Waters nanoEase M/Z Peptide BEH C18 Column, 130Å, 1.7μm, 300μm x 150 mm
Catalog No. 502038954 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The nanoEase M/Z Peptide BEH C18 Column provides optimal performance in nano- to micro-scale liquid chromatographic separation, ensuring high sensitivity and resolution for biomarker discovery and protein identification. It features a 300 μm ID, ultra-low dispersion design, and withstands 15kpsi operating pressure for superior longevity. Utilizing patented BEH Technology with 130Å pore, C18, 1.7 μm particles, it offers a wide usable pH range and ultra-low column bleed, ideal for proteomics.
  • ZenFit connector provides easy, tool-free liquid connections.
  • Ensures low backpressure, wide dynamic range, and long column lifetime.
  • Features a pH range of 1 to 12.
  • Withstands up to 15000 psi (1034 Bar) operating pressure.
  • Spherical particle shape with low silanol activity.
  • QC tested for peptide separation.
Catalog No. 50-203-8954 Supplier Waters Corp Supplier No. 186009259
The nanoEase M/Z Peptide BEH C18 Column provides optimal performance in nano- to micro-scale liquid chromatographic separation, ensuring high sensitivity and resolution for biomarker discovery and protein identification. It features a 300 μm ID, ultra-low dispersion design, and withstands 15kpsi operating pressure for superior longevity. Utilizing patented BEH Technology with 130Å pore, C18, 1.7 μm particles, it offers a wide usable pH range and ultra-low column bleed, ideal for proteomics.
  • ZenFit connector provides easy, tool-free liquid connections.
  • Ensures low backpressure, wide dynamic range, and long column lifetime.
  • Features a pH range of 1 to 12.
  • Withstands up to 15000 psi (1034 Bar) operating pressure.
  • Spherical particle shape with low silanol activity.
  • QC tested for peptide separation.