Learn More
Waters Corp Waters Column, C18, 1.7 μm, 300 Å, 300 μm ID
Supplier: Waters Corp 186009263
These columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, providing high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization.
- 300μm ID columns built with an ultra-low dispersion design.
- Withstand 15,000 psi operating pressure, providing superior longevity and handling pressure fluctuations.
- Contains BEH Technology 300Å pore, C18, 1.7 μm particles.
- Wide-pore (300Å) trifunctionally bonded particle offers widest usable pH range and superior low pH stability.
- Features ultra-low column bleed for analyzing large peptides in proteomics and protein characterization.
- Each batch specifically QC tested using a gradient separation of a tryptic digest of cytochrome c before being QC accepted as a peptide separation column.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.