Learn More
Waters Corp Waters Column, C18, 1.7 μm, 130 Å, 300 μm x 50 mm
Catalog No. 502038952 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Engineered for nano- to micro-scale liquid chromatographic separation, these columns deliver high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. They feature an ultra-low dispersion design, ease of use, and withstand 15kpsi operating pressure for superior longevity.
  • Patented BEH Technology 130Å pore, C18, 1.7 μm particles for wide pH range, low pH stability, and ultra-low column bleed.
  • QC tested for peptide separation; produced in cGMP, ISO 9001-certified facility with chromatographic testing.
  • Individually examined with an intelligent chip for performance and batch analysis.
  • Includes pre-assembled ZenFit Connectors (10-32 threaded port for 1/16 in. O.D. tubing) for fingertight, reusable, 90°C-resistant connections.
  • Ultra-low dispersion design and high pressure resistance enhance lifetime.
Catalog No. 50-203-8952 Supplier Waters Corp Supplier No. 186009257
Engineered for nano- to micro-scale liquid chromatographic separation, these columns deliver high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. They feature an ultra-low dispersion design, ease of use, and withstand 15kpsi operating pressure for superior longevity.
  • Patented BEH Technology 130Å pore, C18, 1.7 μm particles for wide pH range, low pH stability, and ultra-low column bleed.
  • QC tested for peptide separation; produced in cGMP, ISO 9001-certified facility with chromatographic testing.
  • Individually examined with an intelligent chip for performance and batch analysis.
  • Includes pre-assembled ZenFit Connectors (10-32 threaded port for 1/16 in. O.D. tubing) for fingertight, reusable, 90°C-resistant connections.
  • Ultra-low dispersion design and high pressure resistance enhance lifetime.