Learn More
Waters Corp Waters nanoEase M/Z Peptide BEH C18 Column, 300Å, 1.7 μm
Catalog No. 502038824 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
nanoEase M/Z Peptide BEH C18 Columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separations, providing high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. They withstand operating pressures up to 15 kpsi, handling pressure fluctuations effectively for superior longevity.
  • Features BEH Technology 300Å pore, C18, 1.7 μm particles.
  • Equipped with the ZenFit connector, an easy-to-use, fingertight, tool-free liquid connection ensuring reproducible, leak-free connections.
  • Withstands 15kpsi operating pressure and handles pressure fluctuations during injection extremely well, providing superior longevity.
Catalog No. 50-203-8824 Supplier Waters Corp Supplier No. 186008803
nanoEase M/Z Peptide BEH C18 Columns are engineered for optimal performance in nano- to micro-scale liquid chromatographic separations, providing high sensitivity, resolution, and reproducibility for biomarker discovery, protein identification, and characterization. They withstand operating pressures up to 15 kpsi, handling pressure fluctuations effectively for superior longevity.
  • Features BEH Technology 300Å pore, C18, 1.7 μm particles.
  • Equipped with the ZenFit connector, an easy-to-use, fingertight, tool-free liquid connection ensuring reproducible, leak-free connections.
  • Withstands 15kpsi operating pressure and handles pressure fluctuations during injection extremely well, providing superior longevity.