Learn More
Waters Corp nanoEase M/Z Trap Column, Peptide BEH C18, 130Å, 5 μm, 300 μm × 50 mm
Supplier:  Waters Corp 186009268
Encompass_Preferred
These columns offer optimal performance in nano- to micro-scale liquid chromatographic separation, providing high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. They feature an ultra-low dispersion design for ease of use and withstand 15000 psi (1034 Bar) operating pressure, ensuring superior longevity.
  • High sensitivity, resolution, and reproducibility.
  • Ultra-low dispersion design for ease of use.
  • Withstands 15000 psi (1034 Bar) operating pressure for superior longevity.
  • Utilizes BEH Technology 130Å pore, C18, 5μm particles.
  • Offers wide usable pH range (1 pH to 12 pH), superior low pH stability, and ultra-low column bleed.
  • Each batch undergoes specific QC testing for peptide separation.
  • Suggested for Bottom-up Proteomics, Nano-flow, and Trap and Elute.
Catalog No. 50-203-8962