Learn More
Waters Corp nanoEase M/Z Peptide CSH C18 Column, 130Å, 1.7 μm, 150 μm X 150 mm
Catalog No. 502391479 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The nanoEase M/Z Peptide CSH C18 Column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. It withstands pressures up to 15000 psi (1034 Bar) for superior longevity. Featuring a ZenFit connector and CSH Technology, it’s ideal for various LC-MS applications.
  • C18 chemistry with hybrid, spherical particles.
  • pH range of 1 to 10; low silanol activity.
  • 1.7 μm particle and 130 Å pore sizes.
  • ZenFit connector for easy, tool-free connections.
  • CSH Technology for excellent peptide peak shape.
  • QC tested with tryptic digest of cytochrome c.
  • 150 μm ID and 150 mm length.
  • Pre-packed liner streamlines workflow.
  • Versatile for LC-MS, proteomics, and drug discovery.
Catalog No. 50-239-1479 Supplier Waters Corp Supplier No. 186008814
The nanoEase M/Z Peptide CSH C18 Column is engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. It withstands pressures up to 15000 psi (1034 Bar) for superior longevity. Featuring a ZenFit connector and CSH Technology, it’s ideal for various LC-MS applications.
  • C18 chemistry with hybrid, spherical particles.
  • pH range of 1 to 10; low silanol activity.
  • 1.7 μm particle and 130 Å pore sizes.
  • ZenFit connector for easy, tool-free connections.
  • CSH Technology for excellent peptide peak shape.
  • QC tested with tryptic digest of cytochrome c.
  • 150 μm ID and 150 mm length.
  • Pre-packed liner streamlines workflow.
  • Versatile for LC-MS, proteomics, and drug discovery.