Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, CSH C18, 75 μm x 200 mm, 1.7 μm

Catalog No. 502391446 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This nanoEase M/Z Peptide CSH C18 Column is designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility for peptide separations in LC or LC-MS applications, including bottom-up proteomics.

  • Features CSH (Charged Surface Hybrid) particle technology.
  • Withstands 15,000 psi (1,034 Bar) operating pressure.
  • Provides superior longevity and excellent peak shape for peptides.
  • Low silanol activity.
  • Equipped with ZenFit connector for easy, fingertight, tool-free liquid connections.
  • QC tested for peptide separation.
  • Particle size: 1.7 μm.
  • Pore size: 130 Å.
  • Inner diameter: 75 μm.
  • Length: 200 mm.
  • pH range: 1 to 10.
  • Endcapped.

Catalog No. 50-239-1446 Supplier Waters Corp Supplier No. 186008809
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This nanoEase M/Z Peptide CSH C18 Column is designed for optimal performance in nano- to micro-scale liquid chromatographic separation, offering high sensitivity, resolution, and reproducibility for peptide separations in LC or LC-MS applications, including bottom-up proteomics.

  • Features CSH (Charged Surface Hybrid) particle technology.
  • Withstands 15,000 psi (1,034 Bar) operating pressure.
  • Provides superior longevity and excellent peak shape for peptides.
  • Low silanol activity.
  • Equipped with ZenFit connector for easy, fingertight, tool-free liquid connections.
  • QC tested for peptide separation.
  • Particle size: 1.7 μm.
  • Pore size: 130 Å.
  • Inner diameter: 75 μm.
  • Length: 200 mm.
  • pH range: 1 to 10.
  • Endcapped.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.