Learn More
Waters Corp Column, CSH C18, 1.7 μm Particle, 130Å Pore
Catalog No. 502392526 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These columns provide optimal nano- to micro-scale liquid chromatographic separation, ensuring high sensitivity, resolution, and reproducibility for biomarker discovery and protein characterization. They withstand 15,000 psi (1,000 Bar) operating pressure for superior longevity. The ZenFit connector offers easy, fingertight, tool-free, leak-free connections, compatible with V-Detail ports and Rheodyne/Vici valves.
  • Features CSH technology 130Å pore, C18, 1.7 μm particles.
  • Optimal for peptide separations.
  • CSH particles provide excellent peak shape for peptides with FA- or TFA-containing mobile phases.
  • Low dependence on strong ion pairing agents, ideal for LC or LC-MS.
  • Batch QC tested using gradient separation of tryptic digest of cytochrome c.
  • Suggested for bottom-up proteomics, nano-flow, and trap and elute.
Catalog No. 50-239-2526 Supplier Waters Corp Supplier No. 186008812
Add to Cart
Add to Cart
These columns provide optimal nano- to micro-scale liquid chromatographic separation, ensuring high sensitivity, resolution, and reproducibility for biomarker discovery and protein characterization. They withstand 15,000 psi (1,000 Bar) operating pressure for superior longevity. The ZenFit connector offers easy, fingertight, tool-free, leak-free connections, compatible with V-Detail ports and Rheodyne/Vici valves.
  • Features CSH technology 130Å pore, C18, 1.7 μm particles.
  • Optimal for peptide separations.
  • CSH particles provide excellent peak shape for peptides with FA- or TFA-containing mobile phases.
  • Low dependence on strong ion pairing agents, ideal for LC or LC-MS.
  • Batch QC tested using gradient separation of tryptic digest of cytochrome c.
  • Suggested for bottom-up proteomics, nano-flow, and trap and elute.