Learn More
Waters Corp Waters nanoEase M/Z Peptide CSH C18 Column, 130Å, 1.7 μm
Supplier:  Waters Corp 186008807
Encompass_Preferred
The nanoEase M/Z Peptide CSH C18 Column is designed for nano- and micro-scale LC-MS applications requiring high sensitivity. It uses ZenFit Connection Technology for consistent results, high pressure, and temperature stability, enhancing mass spectrometry by improving sampling and ionization efficiency.
  • Re-usable finger-tight fittings with ZenFit connection technology, withstanding up to 15,000 psi and operating at up to 90 °C.
  • 1.7 μm particle size and 130Å pore size.
  • 75 μm × 100 mm dimensions.
  • Enhances mass spectrometry sensitivity by increasing sampling and ionization efficiency, and reducing matrix effects.
  • Prevents poor connections, maintains flow path integrity, and simplifies setup for consistent chromatographic results.
  • Withstands high pressures and temperatures for robust performance.
Catalog No. 50-203-8828