Learn More
Waters Corp nanoEase M/Z Peptide CSH C18 Column, 130Ã…, 1.7 µm, 300 µm X 150 mm, 1/pk;
Supplier:  Waters Corp 186009256
Encompass_Preferred
nanoEase M/Z Peptide CSH C18 Column, 130Å, 1.7 µm, 300 µm X 150 mm, 1/pk;
Catalog No. 50-203-8951