Learn More
Waters Corp Waters Column, nanoEase M/Z Peptide, CSH C18, 1.7 μm, 300 μm ID
Catalog No. 502038949 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The nanoEase M/Z Peptide CSH C18 Column offers optimal performance in nano- to micro-scale liquid chromatographic separation, delivering high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. It withstands 15kpsi operating pressure, ensuring superior longevity, and features patented CSH Technology with 130Å pore, C18, 1.7 μm particles, ideal for LC or LC-MS applications.
  • Optimal performance for nano- to micro-scale liquid chromatographic separation.
  • Withstands 15kpsi operating pressure for superior longevity.
  • Features CSH Technology 130Å pore, C18, 1.7 μm particles.
  • Excellent peptide peak shape; ideal for LC or LC-MS applications.
  • End-capped to minimize MS bleed, improving analysis sensitivity.
Catalog No. 50-203-8949 Supplier Waters Corp Supplier No. 186009254
The nanoEase M/Z Peptide CSH C18 Column offers optimal performance in nano- to micro-scale liquid chromatographic separation, delivering high sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. It withstands 15kpsi operating pressure, ensuring superior longevity, and features patented CSH Technology with 130Å pore, C18, 1.7 μm particles, ideal for LC or LC-MS applications.
  • Optimal performance for nano- to micro-scale liquid chromatographic separation.
  • Withstands 15kpsi operating pressure for superior longevity.
  • Features CSH Technology 130Å pore, C18, 1.7 μm particles.
  • Excellent peptide peak shape; ideal for LC or LC-MS applications.
  • End-capped to minimize MS bleed, improving analysis sensitivity.