Learn More
Waters Corp nanoEase M/Z Peptide CSH C18 Column, 130Å, 1.7 µm, 150 µm X 100 mm, 1/pk
Supplier:  Waters Corp 186008813
Encompass_Preferred
"Waters nanoEase M/Z Columns are designed for optimal performance for nano- to micro scale liquid chromatographic separation for biomarker discovery and protein and peptide identification and characterization. All nanoLC columns are designed for use up to 15 kpsi (1,000 bar) operating pressure and handle pressure fluctuations during injection extremely well and thus provide superior longevity.The columns are equipped with the ZenFit connector, an easy to use, fingertight, tool free liquid connection. This ensures reproducible, leak free connections. The fittings will fit into Waters V-Detail ports as well as into Rheodyne/Vici compatible valves."
Catalog No. NC1868307