Learn More
Waters Corp nanoEase MZ Symmetry C18 Trap Column, 100A, 5 um 180 um x 20mm, 1/pkg;Silica, USP Class:L1 , 2 - 8 Ph Range, Low Silanol Activity, 335 m2/g, End Capped , Spherical , 18 % Carbon Load
Catalog No. 501461986 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
nanoEase MZ Symmetry C18 Trap Column, 100A, 5 um 180 um x 20mm, 1/pkg;Silica, USP Class:L1 , 2 - 8 Ph Range, Low Silanol Activity, 335 m2/g, End Capped , Spherical , 18 % Carbon Load
Catalog No. 50-146-1986 Supplier Waters Corp Supplier No. 186008821
nanoEase MZ Symmetry C18 Trap Column, 100A, 5 um 180 um x 20mm, 1/pkg;Silica, USP Class:L1 , 2 - 8 Ph Range, Low Silanol Activity, 335 m2/g, End Capped , Spherical , 18 % Carbon Load