Learn More
Waters Corp nanoEase MZ Peptide BEH C18 Column, 130Å, 1.7 µm, 75 µm x 250 mm, 1/pkg;Hybrid, USP Class:L1 , 2 - 10 Ph Range, Low Silanol Activity, 185 m2/g, End Capped , Spherical , 18 % Carbon Load
Catalog No. 501461960 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
nanoEase MZ Peptide BEH C18 Column, 130Å, 1.7 µm, 75 µm x 250 mm, 1/pkg;Hybrid, USP Class:L1 , 2 - 10 Ph Range, Low Silanol Activity, 185 m2/g, End Capped , Spherical , 18 % Carbon Load
Catalog No. 50-146-1960 Supplier Waters Corp Supplier No. 186008795
nanoEase MZ Peptide BEH C18 Column, 130Å, 1.7 µm, 75 µm x 250 mm, 1/pkg;Hybrid, USP Class:L1 , 2 - 10 Ph Range, Low Silanol Activity, 185 m2/g, End Capped , Spherical , 18 % Carbon Load