Learn More
Alstem NANOFECT TRANSFECTION RGNT,1ML
SDP
Catalog No. NC1551908
Encompass
NanoFect Transfection Reagent: 1mlNanoFect Transfection Reagent is a unique blend of polymers that delivers more DNA and siRNAs to cells than the leading lipid-based transfection kits, at a lower cost. NanoFect™ is a powerful, broadly applicable transfection reagent for effective and reproducible transfections.
Catalog No. NC1551908 Supplier Alstem Supplier No. NF100
NanoFect Transfection Reagent: 1mlNanoFect Transfection Reagent is a unique blend of polymers that delivers more DNA and siRNAs to cells than the leading lipid-based transfection kits, at a lower cost. NanoFect™ is a powerful, broadly applicable transfection reagent for effective and reproducible transfections.