Learn More
World Precision Instrument NanoFil Injection Holder
SDP
Catalog No. NC0933395
Encompass
Use this holder for direct hand-held injections. The NFINHLD is 18mm long and has a diameter of 9.6mm (there are also two flat spots to 8.6mm on the 0 and 180 degree axis). The cap is 8.1mm diameter.
Catalog No. NC0933395 Supplier World Precision Instrument Supplier No. NFINHLD
Use this holder for direct hand-held injections. The NFINHLD is 18mm long and has a diameter of 9.6mm (there are also two flat spots to 8.6mm on the 0 and 180 degree axis). The cap is 8.1mm diameter.