Learn More
World Precision Instrument NanoFil Needles-NF33-36BL
SDP
Catalog No. 502521062
Encompass
The replaceable needles used with the NanoFil are available with either blunt or beveled tips. The blunt tip is used for injection into soft tissue and when a uniform solution distribution is needed. The beveled style is used for applications that involve the penetration of a tough tissue.
Catalog No. 50-252-1062 Supplier World Precision Instrument Supplier No. NF3336BL
The replaceable needles used with the NanoFil are available with either blunt or beveled tips. The blunt tip is used for injection into soft tissue and when a uniform solution distribution is needed. The beveled style is used for applications that involve the penetration of a tough tissue.