Learn More
World Precision Instrument Sub-Microliter Injection System-NANOFIL
SDP
Catalog No. NC0578818
Encompass
The NanoFilTM sub-microliter injection system boasts zero dead volume. Variability is eliminated with the NanoFilTM  design: our interchangable needles advance directly into the syringe barrel, creating a 1:1 connection with the plunger— a truly gas-tight system built for low-volume sample control.
Our cross-compatible NanoFilTM needles are offered in sizes from 33-36g, blunt or beveled. 
Catalog No. NC0578818 Supplier World Precision Instrument Supplier No. NANOFIL
The NanoFilTM sub-microliter injection system boasts zero dead volume. Variability is eliminated with the NanoFilTM  design: our interchangable needles advance directly into the syringe barrel, creating a 1:1 connection with the plunger— a truly gas-tight system built for low-volume sample control.
Our cross-compatible NanoFilTM needles are offered in sizes from 33-36g, blunt or beveled. 