Learn More
Thomson Instrument Company nano | Filter Vial PTFE 0.45µm, Non-Slit Septum, Blue Cap
SDP
Supplier:  Thomson Instrument Company 15540200
Encompass_Preferred
Thomson nano|Filter Vials offer a low dead volume, allowing filtration with as little as 10 µl of sample with enough remaining filtrate to make a 2 µl injection.Minimal sample loss makes the nano|Filter Vials ideal for critical samples replacing syringe filters, spin columns, autosampler vial, low volume insert, and cap.Simply add the sample and sample buffer, press, and it is ready for the UPLC or HPLC.
•In vial filtration with minimal sample loss, 10 µl sample for a 2 µl injection
•Minimize sample loss by eliminating multiple transfers
•Compatible with standard UHPLC or HPLC autosamplers (12×32 mm)
•Available in 0.2 µm and 0.45 µm pore sizes
•Split and non-split septums available
• Made in the USA
Catalog No. NC0497472