Learn More
Waters Corp NanoFlow/LockSpray Outer Source Service Kit, MS System, Rebuild Kit
Catalog No. 502392022 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This service kit is designed for maintaining and rebuilding a NanoFlow/LockSpray Outer Source. It includes O-rings and fittings specifically for the reference probe and is used in NanoLockSpray systems.
  • Designed for maintaining and rebuilding an outer source.
  • Includes O-rings and fittings for the reference probe.
  • Functions as a rebuild kit.
  • Compatible with MS systems.
  • Replacement is as required.
  • Comes with 1 unit per package.
  • Listed as an optional part.
Catalog No. 50-239-2022 Supplier Waters Corp Supplier No. 700004676
This service kit is designed for maintaining and rebuilding a NanoFlow/LockSpray Outer Source. It includes O-rings and fittings specifically for the reference probe and is used in NanoLockSpray systems.
  • Designed for maintaining and rebuilding an outer source.
  • Includes O-rings and fittings for the reference probe.
  • Functions as a rebuild kit.
  • Compatible with MS systems.
  • Replacement is as required.
  • Comes with 1 unit per package.
  • Listed as an optional part.