Learn More
Waters Corp Waters Nanoflow Probe Tip, Thin Wall, Borosilicate Glass
Catalog No. 50793352 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This nanoflow probe tip features a thin wall design and is constructed from borosilicate glass. It has a 1 mm outer diameter and a 0.78 mm inner diameter, and is coated with 30 nm of gold.
  • Thin wall design
  • Borosilicate glass material
  • 1 mm outer diameter
  • 0.78 mm inner diameter
  • 30 nm gold coating
  • Sold in packs of 10
  • Compatible with Eppendorf Gelloader 1-10
Catalog No. 50-793-352 Supplier Waters Corp Supplier No. M956232AD1S
This nanoflow probe tip features a thin wall design and is constructed from borosilicate glass. It has a 1 mm outer diameter and a 0.78 mm inner diameter, and is coated with 30 nm of gold.
  • Thin wall design
  • Borosilicate glass material
  • 1 mm outer diameter
  • 0.78 mm inner diameter
  • 30 nm gold coating
  • Sold in packs of 10
  • Compatible with Eppendorf Gelloader 1-10