Learn More
Aviva Systems Biology Corp NANOG Antibody - N-terminal region (P100591_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100591_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against NANOG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NANOG. Alternate names: ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-62653