Learn More
Aviva Systems Biology Corp NANOG Antibody - N-terminal region (ARP47988_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47988_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NANOG. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human NANOG. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Goat, Horse, Human, Pig, Sheep
Catalog No. 50-014-59695