Learn More
Medchemexpress LLC NANOG Human His-SU 10ug
SDP
Catalog No. 5000179688
Encompass_Preferred
NANOG Protein Human (His-SUMO) which functions to maintain the undifferentiated state of pluripotent stem cells
Catalog No. 50-001-79688 Supplier Medchemexpress LLC Supplier No. HYP739110UG
NANOG Protein Human (His-SUMO) which functions to maintain the undifferentiated state of pluripotent stem cells