Learn More
ABclonal Technology Nanog Rabbit mAb
SDP
Catalog No. 5000006850
Encompass_Preferred
The protein encoded by this gene is a DNA binding homeobox transcription factor involved in embryonic stem (ES) cell proliferation renewal and pluripotency The encoded protein can block ES cell differentiation and can also autorepress its own expression in differentiating cells Two transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-06850 Supplier ABclonal Technology Supplier No. A22625100UL
Add to Cart
Add to Cart
The protein encoded by this gene is a DNA binding homeobox transcription factor involved in embryonic stem (ES) cell proliferation renewal and pluripotency The encoded protein can block ES cell differentiation and can also autorepress its own expression in differentiating cells Two transcript variants encoding different isoforms have been found for this gene