Learn More
Bioworld Nanogold*- rabbit (NRI) AB, 0.5 mL
Catalog No. 50198350
Encompass_Preferred
Nanogold* shines when it comes to EM. It is the new gold standard. Nanogold* reagents diffuse easily tens of microns into tissues and into permeabilized cells and cell nuclei, to where most colloidal gold probes have difficulty in reaching.Smallest commer
Catalog No. 50-198-350 Supplier Bioworld Supplier No. 203300081
Nanogold* shines when it comes to EM. It is the new gold standard. Nanogold* reagents diffuse easily tens of microns into tissues and into permeabilized cells and cell nuclei, to where most colloidal gold probes have difficulty in reaching.Smallest commer