Learn More
World Precision Instrument NANOLITER2020 Injector Head
SDP
Catalog No. 502521476
Encompass
Simple-to-Use Touchscreen Controller
NANOLITER2020 is the updated version of WPI’s popular NANOLITER2010 Injector that is capable of performing precision injections in the nanoliter range using a variety of glass micropipette sizes. The new model connects directly to the WPI MICRO2T SMARTouch™ controller, which can also be used with WPI’s UMP3 pump, allowing the pumps to be controlled individually or synchronously. The new injector head has a LED indicator which shows proper communication between the pump and controller. NANOLITER2020 offers improved precision by in-depth plunger displacement validation. The system is easier to use and has a chic silver injector head.
Catalog No. 50-252-1476 Supplier World Precision Instrument Supplier No. 300704
Simple-to-Use Touchscreen Controller
NANOLITER2020 is the updated version of WPI’s popular NANOLITER2010 Injector that is capable of performing precision injections in the nanoliter range using a variety of glass micropipette sizes. The new model connects directly to the WPI MICRO2T SMARTouch™ controller, which can also be used with WPI’s UMP3 pump, allowing the pumps to be controlled individually or synchronously. The new injector head has a LED indicator which shows proper communication between the pump and controller. NANOLITER2020 offers improved precision by in-depth plunger displacement validation. The system is easier to use and has a chic silver injector head.