Learn More
Mitegen Clipping Tool Set, Includes 4 Grid Clipping Tools, 1 C-Clip Bottoming Tool & 1 Tool Rack, For Cryo-EM
SDP
Catalog No. NC2220163
Encompass
This Clipping Tool Set by Nanosoft provides essential instruments for precise and efficient grid clipping in cryo-electron microscopy workflows. Designed to streamline your sample preparation, this comprehensive set includes four high-quality grid clipping tools. These tools are engineered to securely handle delicate grids, ensuring accurate placement for subsequent cryo-EM analysis. The set also features one C-clip bottoming tool, which facilitates the proper seating of C-clips onto the clipping tool barrels, minimizing mis-bottoming and preventing loss of small components. For convenient organization and accessibility, one dedicated clipping tool rack is included, keeping your workspace tidy and tools readily available. This complete set enhances precision, efficiency, and reliability during crucial grid preparation steps, making it an indispensable asset for any laboratory engaged in advanced cryo-EM research. Includes 4 Grid Clipping Tools, 1 C-Clip Bottoming Tool & 1 Tool Rack.
This item is not returnable. View return policy
Catalog No. NC2220163 Supplier Mitegen Supplier No. MCEMNSCLTSET
This Clipping Tool Set by Nanosoft provides essential instruments for precise and efficient grid clipping in cryo-electron microscopy workflows. Designed to streamline your sample preparation, this comprehensive set includes four high-quality grid clipping tools. These tools are engineered to securely handle delicate grids, ensuring accurate placement for subsequent cryo-EM analysis. The set also features one C-clip bottoming tool, which facilitates the proper seating of C-clips onto the clipping tool barrels, minimizing mis-bottoming and preventing loss of small components. For convenient organization and accessibility, one dedicated clipping tool rack is included, keeping your workspace tidy and tools readily available. This complete set enhances precision, efficiency, and reliability during crucial grid preparation steps, making it an indispensable asset for any laboratory engaged in advanced cryo-EM research. Includes 4 Grid Clipping Tools, 1 C-Clip Bottoming Tool & 1 Tool Rack.