Learn More
Mitegen Plastic Ethane Condenser by Nanosoft, For One-Handed Liquid Ethane Filling, Interfaces with Vitrobot Spider, Reduces Waste
SDP
Catalog No. NC1816907
Encompass
This Plastic Ethane Condenser by Nanosoft is a custom-designed tool engineered for efficient and ergonomic liquid ethane filling in cryo-electron microscopy workflows. It seamlessly interfaces with the Vitrobot spider, integrating directly into the standard styrofoam dewar set. This innovative design enables true one-handed ethane condensing, allowing the user to focus solely on the liquid ethane level and control the ethane flow rate with precision. By eliminating the need for manual nozzle handling, this condenser significantly simplifies the filling process, enhancing user control and comfort. Furthermore, it helps reduce wasted ethane gas and minimizes the time required to achieve optimal liquid ethane levels in the cup. This leads to more consistent sample vitrification and improved overall workflow efficiency for delicate cryo-EM sample preparation. It is an indispensable accessory for streamlining your cryo-EM operations.
Catalog No. NC1816907 Supplier Mitegen Supplier No. MCEMNSEC01
This Plastic Ethane Condenser by Nanosoft is a custom-designed tool engineered for efficient and ergonomic liquid ethane filling in cryo-electron microscopy workflows. It seamlessly interfaces with the Vitrobot spider, integrating directly into the standard styrofoam dewar set. This innovative design enables true one-handed ethane condensing, allowing the user to focus solely on the liquid ethane level and control the ethane flow rate with precision. By eliminating the need for manual nozzle handling, this condenser significantly simplifies the filling process, enhancing user control and comfort. Furthermore, it helps reduce wasted ethane gas and minimizes the time required to achieve optimal liquid ethane levels in the cup. This leads to more consistent sample vitrification and improved overall workflow efficiency for delicate cryo-EM sample preparation. It is an indispensable accessory for streamlining your cryo-EM operations.