Learn More
Mitegen Reverse Action Autogrid Tweezers by Nanosoft, Notched Tip for Secure Autogrid & Cassette Placement, Prevents Dropping
SDP
Catalog No. NC2171344
Encompass
These Reverse Action Autogrid Tweezers are expertly designed to enhance the precision and security of autogrid manipulation for cryo-electron microscopy workflows. They feature a specialized notched tip geometry that safely cradles autogrids, ensuring they are placed centered in the cassette slot and at the correct depth. The innovative reverse-action handle is a key advantage, as it securely holds the autogrid in its natural, relaxed state. This design significantly simplifies orientation and manipulation steps, ensuring that valuable clipped grids cannot be accidentally dropped during critical procedures. These tweezers make the handling process easier, more reliable, and less stressful. An optional insulating foam can be added to the handles to provide thermal insulation while retaining dexterity, further enhancing user comfort and control during cryogenic transfers. This tool is indispensable for maintaining sample integrity and optimizing your cryo-EM workflow.
This item is not returnable. View return policy
Catalog No. NC2171344 Supplier Mitegen Supplier No. MCEMNSRAAGTW
These Reverse Action Autogrid Tweezers are expertly designed to enhance the precision and security of autogrid manipulation for cryo-electron microscopy workflows. They feature a specialized notched tip geometry that safely cradles autogrids, ensuring they are placed centered in the cassette slot and at the correct depth. The innovative reverse-action handle is a key advantage, as it securely holds the autogrid in its natural, relaxed state. This design significantly simplifies orientation and manipulation steps, ensuring that valuable clipped grids cannot be accidentally dropped during critical procedures. These tweezers make the handling process easier, more reliable, and less stressful. An optional insulating foam can be added to the handles to provide thermal insulation while retaining dexterity, further enhancing user comfort and control during cryogenic transfers. This tool is indispensable for maintaining sample integrity and optimizing your cryo-EM workflow.