Learn More
Mitegen Vitrification Foam Dewar with Foam Ring by Nanosoft, Compatible with Vitrobot & Custom Cryo-EM Sample Prep Systems
SDP
Catalog No. NC2220155
Encompass
This Vitrification Foam Dewar, which includes a complementary foam ring, provides a robust and easy-to-use option for both Vitrobot systems and custom cryo-electron microscopy (cryo-EM) sample preparation platforms. Designed for reliable performance in cryogenic environments, this foam dewar maintains stable temperatures crucial for vitrification and sensitive sample handling. It ensures an optimal environment for preparing delicate samples by securely holding essential metal components such as the ethane cup, grid box ring, and spider. The included foam ring further enhances thermal insulation and helps minimize frost formation, contributing to cleaner sample preparation. This dewar is an indispensable tool for researchers building custom cryo-EM systems or seeking a dependable replacement for their existing Vitrobot setup. It streamlines the cryo-EM workflow, promoting consistency and safeguarding valuable biological specimens during flash-freezing processes.
This item is not returnable. View return policy
Catalog No. NC2220155 Supplier Mitegen Supplier No. MCEMNSVBFD
This Vitrification Foam Dewar, which includes a complementary foam ring, provides a robust and easy-to-use option for both Vitrobot systems and custom cryo-electron microscopy (cryo-EM) sample preparation platforms. Designed for reliable performance in cryogenic environments, this foam dewar maintains stable temperatures crucial for vitrification and sensitive sample handling. It ensures an optimal environment for preparing delicate samples by securely holding essential metal components such as the ethane cup, grid box ring, and spider. The included foam ring further enhances thermal insulation and helps minimize frost formation, contributing to cleaner sample preparation. This dewar is an indispensable tool for researchers building custom cryo-EM systems or seeking a dependable replacement for their existing Vitrobot setup. It streamlines the cryo-EM workflow, promoting consistency and safeguarding valuable biological specimens during flash-freezing processes.