Learn More
Microbiologics™ Nanosphere Verigene™ Gram-Negative Blood Culture Control Panel
SDP
Catalog No. 23004122 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
15 Total Vials (5 vials of each pool)
Catalog No. Quantity
23-004-122 15 Total Vials (5 vials of each pool)
1 options
This item is not returnable. View return policy
Catalog No. 23-004-122 Supplier Microbiologics™ Supplier No. 8181
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains manufacturer recommended inactivated QC microorganisms. Microbiologics™ Nanosphere Verigene™ Gram-Negative Blood Culture Control Panel comes with 5 vials of each pool.
  • Consists of 5 vials of each pool (15 total vials)
  • Features color coordinated labels for easy pool identification
List of strains included in each pool:
  • Pool 4: A. baumannii NCIMB 12457; P. aeruginosa NCTC 10662; E. cloacae NCTC 13464
  • Pool 5: K. pneumoniae NCTC 13438; C. freundii NCTC 9750; E. coli NCTC 13476; K. pneumoniae NCTC 13443; K. pneumoniae NCTC 13440
  • Pool 6: K. oxytoca NCTC 11686; K. pneumoniae NCTC 13442; P. vulgaris NCTC 4636
Specifications
Biosafety Level 1
Format Helix Elite™
Product Type Blood Culture Control Panel
Includes Three pools containing 3 to 4 microorganisms per pool
Quantity 15 Total Vials (5 vials of each pool)
Helix Elite QC Panels and Molecular Standards are FDA Class I exempt unassayed, in vitro diagnostic external controls, and are intended for professional laboratory use only. Results with diagnostic instruments may vary.