Learn More
Microbiologics™ Nanosphere Verigene™ Gram-Positive Blood Culture Control Panel
SDP
Catalog No. 23004119 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
15 Total Vials (5 vials of each pool)
Catalog No. Quantity
23-004-119 15 Total Vials (5 vials of each pool)
1 options
This item is not returnable. View return policy
Catalog No. 23-004-119 Supplier Microbiologics™ Supplier No. 8180
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains manufacturer recommended inactivated QC microorganisms. Microbiologics™ Nanosphere Verigene™ Gram-Positive Blood Culture Control Panel comes with 5 vials of each pool.
  • Consists of 5 vials of each pool (15 total vials)
  • Features color coordinated labels for easy pool identification
List of strains included in each pool:
  • Pool 1: S. epidermidis NCIMB 8853; S. lugdunensis NCTC 7990; E. faecium NCTC 12204; L. monocytogenes NCTC 10890
  • Pool 2: S. pyogenes NCIMB 13285; S. agalactiae NCTC 8017; S. anginosus NCTC 10713
  • Pool 3: S. aureus NCTC 12493; E. faecalis NCTC 13379; S. pneumoniae NCIMB 13286
Specifications
Biosafety Level 1
Format Helix Elite™
Product Type Blood Culture Control Panel
Includes Three pools containing 3 to 4 microorganisms per pool
Quantity 15 Total Vials (5 vials of each pool)
Helix Elite QC Panels and Molecular Standards are FDA Class I exempt unassayed, in vitro diagnostic external controls, and are intended for professional laboratory use only. Results with diagnostic instruments may vary.