Learn More
Cytiva (Formerly Pall Lab) Nanosep Centrifugal Devices with Omega™ Membrane - 100K, clear (100/pkg)
Supplier:  Cytiva OD100C34
Encompass
wwPTFE is an all-purpose, hydrophilic membrane that offers low protein binding and low levels of UV-absorbing extractables This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0388807