Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Nanotechnology: Ferrofluids
GREENER_CHOICESCI_ED

Catalog No. s96596 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S96596 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S96596 Supplier Innovating Science™ Supplier No. IS2800
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Material for 15 groups: 1 g Ferrous Chloride; 22g Ferric Chloride; 20mL Hydrochloric Acid, 10N; 25 mL Ammonium Hydroxide (2); 20g Tetramethylammonium Hydroxide; Pipets (15); Petri dishes (15); teacher's guide; and student copy-masters

Understand the definition of nanotechnology as it applies in current usage.

  • Learn about one nanomaterial, ferrofluid, and some of its applications in modern technology
  • Create magnetite nanoparticles through a precipitation reaction
  • Employ a surfactant to create a colloidal suspension of magnetite nanoparticles (ferrofluid) Examine the response of ferrofluid upon exposure to a magnetic field

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Learn nanomaterial, ferrofluid and some of its applications
Includes Material for 15 groups: 1 g ferrous chloride, 22g ferric chloride, 20ml hydrochloric acid, 10n, 25 mL ammonium hydroxide (2), 20g tetramethylammonium hydroxide, pipets (15), petri dishes (15), teacher's guide, and student copy-masters
Quantity 1
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 15 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.