Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck Nanotrap Enhancement Reagent 2
SDP

Catalog No. 23285975 Shop All Streck Laboratories Products
Change view
Click to view available options
Capacity:
10 mL
30 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity
23-285-975 30 mL
23-285-974 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-285-975 Supplier Streck Supplier No. 250073
This item is not returnable. View return policy

Use of Nanotrap Enhancement Reagents with Nanotrap Microbiome Particles improves the binding of microbes in wastewater samples, further enhancing downstream detection of nucleic acids.

  • Utilize affinity capture and enrichment to attract and bind low abundance viruses.
  • Can be used with clinical swab samples, saliva, urine, wastewater, food wash and blackwater.
  • Compatible with PCR, sequencing or immunoassay analysis methods.
  • Designed to capture and concentrate a variety of microbes.
  • Increase sensitivity of existing SARS-CoV-2 assay by capturing and concentrating whole virus.
  • Nanotrap Enhancement Reagents improve microbe detection in wastewater samples by at least 1-2 Ct values.

Specifications

Capacity 30 mL
Isolation Technology Magnetic Particles
Sample Type Wastewater
Sequencing Type RT-PCR and Digital PCR Sequencing
Product Type Enhancement Reagent 2
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.