Learn More
Description
- Utilize affinity capture and enrichment to attract and bind low abundance viruses.
- Can be used with clinical swab samples, saliva, urine, wastewater, food wash and blackwater.
- Compatible with PCR, sequencing or immunoassay analysis methods.
- Designed to capture and concentrate a variety of microbes.
- Increase sensitivity of existing SARS-CoV-2 assay by capturing and concentrating whole virus.
- Nanotrap Enhancement Reagents improve microbe detection in wastewater samples by at least 1-2 Ct values.
Specifications
Specifications
| Capacity | 10 mL |
| Isolation Technology | Magnetic Particles |
| Sample Type | Wastewater |
| Sequencing Type | RT-PCR and Digital PCR Sequencing |
| Product Type | Microbiome B Particles |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
