Learn More
Thermo Scientific nanoViper™ Fitting Systems for Vanquish™ Neo UHPLC SystemCatalog number: 6250.5135, nanoViper 0.010 x 350 mm (1500 bar)
Catalog No. NC2885195 Shop All Thermo Scientific Products
Encompass
Description- nanoViper 0.010 x 350 mm (1500 bar)For Use With (Application)-Fluidic Connection for Low-Flow UHPLC SystemsFor Use With (Equipment)- All HPLC and UHPLC SystemsMateria- Shielded Fused SilicaMax. Pressure- 1500 barProduct Line- nanoViperType- TubingLength (Metric)- 350 mmDiameter (Metric) Inner- 10 μmUnit Size Each
Catalog No. NC2885195 Supplier Thermo Scientific Supplier No. 6250.5135
Description- nanoViper 0.010 x 350 mm (1500 bar)For Use With (Application)-Fluidic Connection for Low-Flow UHPLC SystemsFor Use With (Equipment)- All HPLC and UHPLC SystemsMateria- Shielded Fused SilicaMax. Pressure- 1500 barProduct Line- nanoViperType- TubingLength (Metric)- 350 mmDiameter (Metric) Inner- 10 μmUnit Size Each