Learn More
Thermo Scientific nanoViper capillary with dimensions 0.020 mm x 550 mm with a 1500 bar tolerance
Catalog No. NC2228879 Shop All Thermo Scientific Products
Encompass
nanoViper capillary with dimensions 0.020 mm x 550 mm with a 1500 bar tolerance for high performance liquid chromatography applications (Vanquish Neo)
This item is not returnable. View return policy
Catalog No. NC2228879 Supplier Thermo Scientific Supplier No. 6250.5260
nanoViper capillary with dimensions 0.020 mm x 550 mm with a 1500 bar tolerance for high performance liquid chromatography applications (Vanquish Neo)