Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific NANOVIPER 0.050 X 350 MM

Catalog No. NC2464662 Shop All Thermo Scientific Products
Encompass

nanoViper 0.050 x 350 mm (

Catalog No. NC2464662 Supplier Thermo Scientific Supplier No. 6250.5540
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

nanoViper 0.050 x 350 mm (

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.