Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Catalog No. 501347942 Shop All Thermo Scientific Products
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies - Preassembled and finger-tightened for easy tool-free instrument setup and configuration - High-pressure tested for UHPLC compatibility up to 1200 bar (17400 psi)
This item is not returnable. View return policy
Catalog No. 50-134-7942 Supplier Thermo Scientific Supplier No. 6041.5813
- Virtually zero-dead-volume connections ensure optimal separation efficiencies - Preassembled and finger-tightened for easy tool-free instrument setup and configuration - High-pressure tested for UHPLC compatibility up to 1200 bar (17400 psi)