Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5815
Encompass
Virtually zero-dead-volume connections,Preassembled and finger-tightened,High-pressure tested up to 1200 bar (17400 psi),Tool-free instrument setup and configuration
Catalog No. 50-134-7944