Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier: Thermo Scientific 6041.5817
- Virtually zero-dead-volume connections ensure optimal separation efficiencies- Preassembled and finger-tightened for easy tool-free instrument setup and configuration- Prevents overtightening- High-pressure tested for UHPLC compatibility up to 1200 bar (17400 psi)