Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5823
Encompass
Virtually zero-dead-volume connections; Preassembled and finger-tightened; High-pressure tested up to 1200 bar (17400 psi); Easy tool-free instrument setup and configuration; Prevents overtightening; Suitable for temperatures up to 120C; Compatible with valves and columns from virtually any manufacturer; Available in different materials, lengths, and inner diameters
Catalog No. 50-134-7952