Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5818
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies- Preassembled and finger-tightened nanoViper fingertight fittings enable easy tool-free instrument setup and configuration and prevent overtightening- High-pressure tested to provide UHPLC compatibility up to 1200 bar (17400 psi)
Catalog No. 50-134-7947