Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Supplier:  Thermo Scientific 6041.5127
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies - Preassembled and finger-tightened for easy tool-free instrument setup and configuration - High-pressure tested for UHPLC compatibility up to 1200 bar (17400 psi) - Ferrule-free tightening design - Operates at pressures up to 1200 bar - Suitable for temperatures up to 120C - Easy to Install with a fingertight design - No tools required - Flexible and compatible with valves and columns from virtually any manufacturer
Catalog No. 50-134-7905