Learn More
Thermo Scientific nanoViper Fingertight Fittings | | 1 EA
Catalog No. 501347900 Shop All Thermo Scientific Products
Encompass
- Virtually zero-dead-volume connections ensure optimal separation efficiencies- Preassembled and finger-tightened nanoViper fingertight fittings enable easy tool-free instrument setup- High-pressure tested to provide UHPLC compatibility up to 1200 bar (17400 psi)- Ferrule-free tightening design- Works with operating pressures up to 1200 bar- Suitable for temperatures up to 120C- Fingertight design- Flexible PEEK sheathed fused silica capillaries, 1/32 in. OD- No tools required- Eliminates assembly of PEEK sleeve fittings- Compatible with valves and columns from virtually any manufacturer- Fits narrow connections such as 10-port valves- Available in different materials, lengths, and inner diameters
Catalog No. 50-134-7900 Supplier Thermo Scientific Supplier No. 6041.5122
- Virtually zero-dead-volume connections ensure optimal separation efficiencies- Preassembled and finger-tightened nanoViper fingertight fittings enable easy tool-free instrument setup- High-pressure tested to provide UHPLC compatibility up to 1200 bar (17400 psi)- Ferrule-free tightening design- Works with operating pressures up to 1200 bar- Suitable for temperatures up to 120C- Fingertight design- Flexible PEEK sheathed fused silica capillaries, 1/32 in. OD- No tools required- Eliminates assembly of PEEK sleeve fittings- Compatible with valves and columns from virtually any manufacturer- Fits narrow connections such as 10-port valves- Available in different materials, lengths, and inner diameters